tert-Butyl-3-(3,3-difluoropyrrolidin-1-yl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115371-100MG 115371-250MG 115371-500MG
102895-822EA 824.62 USD
102895-822 102895-824 102895-826
tert-Butyl-3-(3,3-difluoropyrrolidin-1-yl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(3,3-difluoropyrrolidin-1-yl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₀F₂N₂O₂
MW: 262.3 g/mol
MDL Number: MFCD22123280
CAS Number: 1257294-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR