tert-Butyl-3-(4,4-difluoro-1-piperidyl)azetidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115370-100MG
115370-250MG
115370-500MG
102895-816EA
824.62
USD
102895-816
102895-818
102895-820
tert-Butyl-3-(4,4-difluoro-1-piperidyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(4,4-difluoro-1-piperidyl)azetidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₃H₂₂F₂N₂O₂ MW: 276.33 g/mol |
MDL Number:
MFCD22123273 CAS Number: 1257293-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...