tert-Butyl-3-(4-chlorobutanoylamino)azetidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115368-100MG
115368-250MG
115368-500MG
102895-804EA
824.62
USD
102895-804
102895-806
102895-808
tert-Butyl-3-(4-chlorobutanoylamino)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(4-chlorobutanoylamino)azetidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₂H₂₁ClN₂O₃ MW: 276.77 g/mol |
MDL Number:
MFCD22123278 CAS Number: 1257294-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...