tert-Butyl-5-(benzyloxycarbonylamino)-3-fluoro-5,6-dihydropyridine-1(2H)-carboxylate ≥95%
Supplier: Matrix Scientific
115367-500MG
115367-1G
102895-800EA
3021.54
USD
102895-800
102895-802
tert-Butyl-5-(benzyloxycarbonylamino)-3-fluoro-5,6-dihydropyridine-1(2H)-carboxylate ≥95%
tert-Butyl-5-(benzyloxycarbonylamino)-3-fluoro-5,6-dihydropyridine-1(2H)-carboxylate
Hazard: Irritant
Formula:
C₁₈H₂₃FN₂O₄ MW: 350.39 g/mol |
MDL Number:
MFCD21601917 CAS Number: 1356342-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...