1-(Azetidin-3-yl)-4-methylpiperidin-4-ol ≥95%

Supplier: Matrix Scientific

115364-100MG 115364-250MG 115364-500MG
102895-788EA 824.62 USD
102895-788 102895-790 102895-792
1-(Azetidin-3-yl)-4-methylpiperidin-4-ol ≥95%
1-(Azetidin-3-yl)-4-methylpiperidin-4-ol

Hazard: Irritant

Formula: C₉H₁₈N₂O
MDL Number: MFCD22123276
CAS Number: 1257293-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR