1-(Azetidin-3-yl)-3-fluoroazetidine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-1,3'-biazetidine

115359-250MG 115359-500MG 115359-1G
102895-710EA 963.08 USD
102895-710 102895-712 102895-714
1-(Azetidin-3-yl)-3-fluoroazetidine ≥95%
1-(Azetidin-3-yl)-3-fluoroazetidine

Hazard: Irritant

Formula: C₆H₁₁FN₂
MW: 130.17 g/mol
MDL Number: MFCD22123282
CAS Number: 1257293-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR