Methyl-4-oxotetrahydrofuran-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-oxotetrahydrofuran-3-carboxylate
115357-500MG
115357-1G
102896-096EA
893.85
USD
102896-096
102896-098
Methyl-4-oxotetrahydrofuran-3-carboxylate ≥95%
Methyl-4-oxotetrahydrofuran-3-carboxylate
Hazard: Irritant
Formula:
C₆H₈O₄ MW: 144.13 g/mol |
MDL Number:
MFCD19707049 CAS Number: 57595-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...