4-(Oxetan-3-yl)piperidine oxalate ≥95%

Supplier: Matrix Scientific

115356-500MG 115356-100MG 115356-250MG
102896-094EA 1820 USD
102896-094 102899-288 102899-290
4-(Oxetan-3-yl)piperidine oxalate ≥95%
4-(Oxetan-3-yl)piperidine oxalate

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MDL Number: MFCD22421728
CAS Number: 1394319-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR