3-Methyloxetan-2-one ≥96%

Supplier: Matrix Scientific

115355-100MG 115355-250MG 115355-500MG
102899-282EA 769.23 USD
102899-282 102899-284 102899-286
3-Methyloxetan-2-one ≥96%
3-Methyloxetan-2-one

Hazard: Irritant

Formula: C₄H₆O₂
MDL Number: MFCD22055146
CAS Number: 1823-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR