Isoquinolin-6-ylboronic acid hydrochloride ≥97%
Supplier: Matrix Scientific
115353-1G
115353-5G
102899-272EA
880
USD
102899-272
102899-274
Isoquinolin-6-ylboronic acid hydrochloride ≥97%
Isoquinolin-6-ylboronic acid hydrochloride
Hazard: Irritant
Formula:
C₉H₉BClNO₂ |
MDL Number:
MFCD18383561 CAS Number: 1236031-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...