Ethyl-1-boc-4-ethyl-4-piperidine carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 1-Boc-4-ethyl-4-piperidine carboxylate
115351-1G
115351-5G
102896-090EA
413.85
USD
102896-090
102896-092
Ethyl-1-boc-4-ethyl-4-piperidine carboxylate ≥97%
Ethyl-1-boc-4-ethyl-4-piperidine carboxylate
Hazard: Irritant
Formula:
C₁₅H₂₇NO₄ MW: 285.39 g/mol |
MDL Number:
MFCD10565657 CAS Number: 188792-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...