m-Carborane-1,7-dicarboxylic acid ≥95%

Supplier: Matrix Scientific

115347-250MG 115347-500MG 115347-1G
102896-012EA 880 USD
102896-012 102896-014 102896-016
m-Carborane-1,7-dicarboxylic acid ≥95%
m-Carborane-1,7-dicarboxylic acid

Hazard: Irritant

Formula: C₄H₂B₁₀O₄
MDL Number: MFCD00157045
CAS Number: 50571-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR