2-(p-Tolylsulfonyl)-6-thia-2-azaspiro[2.5]octane ≥95%

Supplier: Matrix Scientific

115343-250MG 115343-500MG 115343-1G
102895-688EA 1318.46 USD
102895-688 102895-690 102895-692
2-(p-Tolylsulfonyl)-6-thia-2-azaspiro[2.5]octane ≥95%
2-(p-Tolylsulfonyl)-6-thia-2-azaspiro[2.5]octane

Hazard: Irritant

Formula: C₁₃H₁₇NO₂S₂
MDL Number: MFCD22123258
CAS Number: 1207754-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR