2-(p-Tolylsulfonyl)-6-oxa-2-azaspiro[2.5]octane ≥95%
Supplier: Matrix Scientific
115342-250MG
115342-500MG
102895-684EA
1318.46
USD
102895-684
102895-686
2-(p-Tolylsulfonyl)-6-oxa-2-azaspiro[2.5]octane ≥95%
2-(p-Tolylsulfonyl)-6-oxa-2-azaspiro[2.5]octane
Hazard: Irritant
Formula:
C₁₃H₁₇NO₃S |
MDL Number:
MFCD22123257 CAS Number: 1207754-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...