Thiophene-2-carboxaldehyde diethylacetal ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(Diethoxymethyl)thiophene
115341-5G
115341-25G
102895-680EA
166.15
USD
102895-680
102895-682
Thiophene-2-carboxaldehyde diethylacetal ≥97%
Thiophene-2-carboxaldehyde diethylacetal
Hazard: Irritant
Formula:
C₉H₁₄O₂S MW: 186.27 g/mol |
MDL Number:
MFCD11109382 CAS Number: 13959-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...