3-Methyloxathiazolidine-2,2-dioxide ≥95%

Supplier: Matrix Scientific

115339-100MG 115339-250MG 115339-500MG
102895-572EA 604.62 USD
102895-572 102895-574 102895-576
3-Methyloxathiazolidine-2,2-dioxide ≥95%
3-Methyloxathiazolidine-2,2-dioxide

Hazard: Irritant

Formula: C₃H₇NO₃S
MDL Number: MFCD22123263
CAS Number: 1201897-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR