1-(N-Methoxycarbonyl)indole-2-boronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (1-(Methoxycarbonyl)-1H-indol-2-yl)boronic acid

115337-1G 115337-5G 115337-25G
102899-258EA 330.77 USD
102899-258 102899-260 102899-262
1-(N-Methoxycarbonyl)indole-2-boronic acid ≥98%
1-(N-Methoxycarbonyl)indole-2-boronic acid

Hazard: Irritant

Formula: C₁₀H₁₀BNO₄
MW: 219.01 g/mol
MDL Number: MFCD22123264
CAS Number: 1001162-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR