S-(1,1-Dioxothiolan-3-yl)ethanethioate ≥95%

Supplier: Matrix Scientific

115335-500MG 115335-1G
102899-244EA 769.23 USD
102899-244 102899-246
S-(1,1-Dioxothiolan-3-yl)ethanethioate ≥95%
S-(1,1-Dioxothiolan-3-yl)ethanethioate

Hazard: Irritant

Formula: C₆H₁₀O₃S₂
MW: 194.27 g/mol
MDL Number: MFCD00195994
CAS Number: 201990-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR