tert-Butyl-4,4-dimethyl-2,2-dioxooxathiazolidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

115333-500MG 115333-1G 115333-5G
102899-234EA 769.23 USD
102899-234 102899-236 102899-238
tert-Butyl-4,4-dimethyl-2,2-dioxooxathiazolidine-3-carboxylate ≥95%
tert-Butyl-4,4-dimethyl-2,2-dioxooxathiazolidine-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₇NO₅S
MDL Number: MFCD22123261
CAS Number: 454248-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR