3-Azidopropylamine ≥95%
Supplier: Matrix Scientific
115329-100MG
115329-250MG
115329-500MG
102895-560EA
495.38
USD
102895-560
102895-562
102895-564
3-Azidopropylamine ≥95%
3-Azidopropylamine
Hazard: Flammable
Formula:
C₃H₈N₄ Flash Pt: 60°C |
MDL Number:
MFCD11046568 CAS Number: 88192-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...