tert-Butyl-1,7-diazaspiro[4.5]decane-1-carboxylate ≥95%

Supplier: Matrix Scientific

115322-500MG 115322-1G 115322-250MG
102899-110EA 1615.38 USD
102899-110 102899-112 102899-228
tert-Butyl-1,7-diazaspiro[4.5]decane-1-carboxylate ≥95%
tert-Butyl-1,7-diazaspiro[4.5]decane-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
MDL Number: MFCD11223544
CAS Number: 960294-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR