9-Boc-3,5-dioxo-2,9-diazaspiro[5.5]undecane ≥95%
Supplier: Matrix Scientific
115320-1G
115320-500MG
115320-5G
102895-552EA
2016.92
USD
102895-552
102896-006
102899-220
9-Boc-3,5-dioxo-2,9-diazaspiro[5.5]undecane ≥95%
9-Boc-3,5-dioxo-2,9-diazaspiro[5.5]undecane
Hazard: Irritant
Formula:
C₁₄H₂₂N₂O₄ MW: 282.34 g/mol |
MDL Number:
MFCD20926184 CAS Number: 1245917-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...