2-Boc-7,9-dioxo-2,6-diazaspiro[3.5]nonane ≥95%
Supplier: Matrix Scientific
115319-500MG
115319-1G
115319-5G
102896-000EA
2692.31
USD
102896-000
102896-002
102896-004
2-Boc-7,9-dioxo-2,6-diazaspiro[3.5]nonane ≥95%
2-Boc-7,9-dioxo-2,6-diazaspiro[3.5]nonane
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₄ MW: 254.29 g/mol |
MDL Number:
MFCD13184430 CAS Number: 1105664-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...