5-(Azidomethyl)-3,3-difluorodihydro-2(3H)-furanone ≥95%

Supplier: Matrix Scientific

115309-500MG 115309-1G
102895-984EA 4393.85 USD
102895-984 102895-986
5-(Azidomethyl)-3,3-difluorodihydro-2(3H)-furanone ≥95%
5-(Azidomethyl)-3,3-difluorodihydro-2(3H)-furanone

Hazard: Irritant

Formula: C₅H₅F₂N₃O₂
CAS Number: 189136-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR