N-(Tetrahydro-4-phenyl-2H-pyran-4-yl)acetamide ≥95%
Supplier: Matrix Scientific
115305-500MG
115305-1G
102895-906EA
1044.62
USD
102895-906
102895-908
N-(Tetrahydro-4-phenyl-2H-pyran-4-yl)acetamide ≥95%
N-(Tetrahydro-4-phenyl-2H-pyran-4-yl)acetamide
Hazard: Irritant
Formula:
C₁₃H₁₉NO₂ MW: 219.284 g/mol |
MDL Number:
MFCD21641995 CAS Number: 17380-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...