(S)-Pyrrolidin-2-ylmethanol hydrochloride ≥95%

Supplier: Matrix Scientific

115304-1G 115304-5G 115304-10G
102894-892EA 193.85 USD
102894-892 102894-894 102894-896
(S)-Pyrrolidin-2-ylmethanol hydrochloride ≥95%
(S)-Pyrrolidin-2-ylmethanol hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD19443980
CAS Number: 79802-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR