(S)-(+)-Pyrrolidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(+)-Pyrrolidine-3-carboxylic Acid

115302-500MG 115302-1G
102894-884EA 989.23 USD
102894-884 102894-886
(S)-(+)-Pyrrolidine-3-carboxylic acid ≥95%
(S)-(+)-Pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD06738463
CAS Number: 72580-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR