tert-Butyl-N-[(6-chloropyrimidin-4-yl)methyl]carbamate ≥95%
Supplier: Matrix Scientific
115299-250MG
115299-500MG
115299-1G
102895-896EA
1318.46
USD
102895-896
102895-898
102895-900
tert-Butyl-N-[(6-chloropyrimidin-4-yl)methyl]carbamate ≥95%
tert-Butyl-N-[(6-chloropyrimidin-4-yl)methyl]carbamate
Hazard: Irritant
Formula:
C₁₀H₁₄ClN₃O₂ MW: 243.69 g/mol |
MDL Number:
MFCD22415185 CAS Number: 1389264-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...