6-(Aminomethyl)pyrimidin-4-amine hydrochloride 95

Supplier: Matrix Scientific

115298-250MG 115298-500MG 115298-1G
102899-214EA 1318.46 USD
102899-214 102899-216 102899-218
6-(Aminomethyl)pyrimidin-4-amine hydrochloride 95
6-(Aminomethyl)pyrimidin-4-amine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₄
MW: 160.61 g/mol
MDL Number: MFCD22199240
CAS Number: 1404373-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR