tert-Butyl-3-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115297-1G
115297-5G
102899-210EA
330.77
USD
102899-210
102899-212
tert-Butyl-3-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-3-(2-ethoxy-2-oxoethyl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₂₅NO₄ MW: 271.36 g/mol |
MDL Number:
MFCD08275144 CAS Number: 384830-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...