tert-Butyl-4-acetamidopiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115296-1G 115296-5G
102899-206EA 330.77 USD
102899-206 102899-208
tert-Butyl-4-acetamidopiperidine-1-carboxylate ≥95%
tert-Butyl-4-acetamidopiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₃
MW: 242.32 g/mol
MDL Number: MFCD09908271
CAS Number: 1093759-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR