5-Bromo-1H-pyrazolo[3,4-b]pyrazin-3-amine ≥95%

Supplier: Matrix Scientific

115295-500MG 115295-1G 115295-5G
102899-200EA 963.08 USD
102899-200 102899-202 102899-204
5-Bromo-1H-pyrazolo[3,4-b]pyrazin-3-amine ≥95%
5-Bromo-1H-pyrazolo[3,4-b]pyrazin-3-amine

Hazard: Irritant

Formula: C₅H₄BrN₅
MDL Number: MFCD17016134
CAS Number: 1242336-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR