4-Bromo-1H-indazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
115294-250MG
115294-500MG
115294-1G
102899-194EA
769.23
USD
102899-194
102899-196
102899-198
4-Bromo-1H-indazole-3-carboxylic acid ≥95%
4-Bromo-1H-indazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅BrN₂O₂ |
MDL Number:
MFCD07367963 CAS Number: 885521-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...