1-Carbobenzoxy-3-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

115293-1G 115293-5G
102899-190EA 330.77 USD
102899-190 102899-192
1-Carbobenzoxy-3-piperidinecarboxylic acid ≥95%
1-Carbobenzoxy-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MDL Number: MFCD02931282
CAS Number: 78190-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR