1-(tert-Butoxycarbonyl)-4-hydroxypiperidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

115292-250MG 115292-500MG
102899-186EA 660 USD
102899-186 102899-188
1-(tert-Butoxycarbonyl)-4-hydroxypiperidine-2-carboxylic acid ≥95%
1-(tert-Butoxycarbonyl)-4-hydroxypiperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
MDL Number: MFCD01861765
CAS Number: 917835-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR