(4-Aminotetrahydro-2H-pyran-4-yl)methanol hydrochloride ≥95%

Supplier: Matrix Scientific

115291-250MG 115291-500MG 115291-1G
102899-180EA 440 USD
102899-180 102899-182 102899-184
(4-Aminotetrahydro-2H-pyran-4-yl)methanol hydrochloride ≥95%
(4-Aminotetrahydro-2H-pyran-4-yl)methanol hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO₂
MDL Number: MFCD22422274
CAS Number: 1404373-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR