4-Benzylpiperazin-1-amine hydrochloride ≥95%

Supplier: Matrix Scientific

115289-1G 115289-5G
102895-888EA 769.23 USD
102895-888 102895-890
4-Benzylpiperazin-1-amine hydrochloride ≥95%
4-Benzylpiperazin-1-amine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD23152700
CAS Number: 40675-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR