2-(Methoxycarbonylamino)-2-(oxetan-3-yl)acetic acid hemioxalate ≥96%

Supplier: Matrix Scientific

115288-100MG 115288-250MG 115288-500MG
102895-882EA 715.38 USD
102895-882 102895-884 102895-886
2-(Methoxycarbonylamino)-2-(oxetan-3-yl)acetic acid hemioxalate ≥96%
2-(Methoxycarbonylamino)-2-(oxetan-3-yl)acetic acid hemioxalate

Hazard: Irritant

Formula: C₆H₁₁NO₃
CAS Number: 394653-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR