1-Methyl-4-(tributylstannyl)imidazole ≥70%

Supplier: Matrix Scientific

115287-500MG 115287-1G 115287-5G
102895-876EA 166.15 USD
102895-876 102895-878 102895-880
1-Methyl-4-(tributylstannyl)imidazole ≥70%
1-Methyl-4-(tributylstannyl)imidazole

Hazard: Irritant

Formula: C₁₆H₃₂N₂Sn
MDL Number: MFCD13193061
CAS Number: 446285-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR