3-Thietaneamine hydrochloride 95

Supplier: Matrix Scientific

115286-500MG 115286-1G 115286-5G
102895-870EA 1238.46 USD
102895-870 102895-872 102895-874
3-Thietaneamine hydrochloride 95
3-Thietaneamine hydrochloride

Hazard: Irritant

Formula: C₃H₈BrNS
MW: 170.08 g/mol
CAS Number: 943437-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR