2-Oxa-6-azaspiro[3.5]nonane oxalate ≥96%

Supplier: Matrix Scientific

115285-100MG 115285-250MG 115285-1G
102895-774EA 495.38 USD
102895-774 102895-776 102895-778
2-Oxa-6-azaspiro[3.5]nonane oxalate ≥96%
2-Oxa-6-azaspiro[3.5]nonane oxalate

Hazard: Irritant

Formula: C₇H₁₃NO
MW: 127.19 g/mol
MDL Number: MFCD20922916
CAS Number: 1366396-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR