1-Boc-3-fluoro-4-pyrrolidinone ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate, 4-Fluoropyrrolidin-3-one, N-BOC protected

115283-250MG 115283-500MG 115283-1G
102895-768EA 1813.85 USD
102895-768 102895-770 102895-772
1-Boc-3-fluoro-4-pyrrolidinone ≥97%
1-Boc-3-fluoro-4-pyrrolidinone

Hazard: Irritant

Formula: C₉H₁₄FNO₃
MW: 203.213 g/mol
MDL Number: MFCD08274459
CAS Number: 845894-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR