1-(Thiazol-2-yl)ethanamine hydrochloride ≥95%
Supplier: Matrix Scientific
115282-1G
115282-5G
102895-764EA
1018.46
USD
102895-764
102895-766
1-(Thiazol-2-yl)ethanamine hydrochloride ≥95%
1-(Thiazol-2-yl)ethanamine hydrochloride
Hazard: Irritant
Formula:
C₅H₉ClN₂S |
MDL Number:
MFCD11846195 CAS Number: 947662-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...