2-Chloro-1-nitro-3-(trifluoromethyl)benzene ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-3-nitrobenzotrifluoride
115274-1G
115274-5G
102894-520EA
112.31
USD
102894-520
102894-522
2-Chloro-1-nitro-3-(trifluoromethyl)benzene ≥95%
2-Chloro-1-nitro-3-(trifluoromethyl)benzene
Hazard: Irritant
Formula:
C₇H₃ClF₃NO₂ Melting Pt: 25…30 °C Flash Pt: |
MDL Number:
MFCD14525509 CAS Number: 39974-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...