Methyl ester-N-(6,8-difluoro-2H-1-benzopyran-3-yl)carbamic acid 95

Supplier: Matrix Scientific

115263-500MG 115263-1G
102893-404EA 1793.85 USD
102893-404 102895-744
Methyl ester-N-(6,8-difluoro-2H-1-benzopyran-3-yl)carbamic acid 95
Methyl ester-N-(6,8-difluoro-2H-1-benzopyran-3-yl)carbamic acid

Hazard: Irritant

Formula: C₁₁H₉F₂NO₃
MW: 241.2 g/mol
MDL Number: MFCD20528372
CAS Number: 1034001-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR