2-(Acetylamino)-3-(4-nitrophenyl)-2-propenoic acid ≥95%
Supplier: Matrix Scientific
115261-500MG
115261-1G
102893-396EA
350.77
USD
102893-396
102893-398
2-(Acetylamino)-3-(4-nitrophenyl)-2-propenoic acid ≥95%
2-(Acetylamino)-3-(4-nitrophenyl)-2-propenoic acid
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₅ |
MDL Number:
MFCD23135432 CAS Number: 70973-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...