Benzyl (2S)-2 amino-3-(thiophene-2-yl)propanoate ≥95%

Supplier: Matrix Scientific

115257-500MG 115257-1G
102893-380EA 804.62 USD
102893-380 102893-382
Benzyl (2S)-2 amino-3-(thiophene-2-yl)propanoate ≥95%
Benzyl (2S)-2 amino-3-(thiophene-2-yl)propanoate

Hazard: Irritant

Formula: C₁₄H₁₅NO₂S
MDL Number: MFCD28023549
CAS Number: 755733-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR