2-(Acetylamino)-3-(2-thienyl)-2-propenoic acid ≥95%

Supplier: Matrix Scientific

115256-500MG 115256-1G
102893-376EA 412.31 USD
102893-376 102893-378
2-(Acetylamino)-3-(2-thienyl)-2-propenoic acid ≥95%
2-(Acetylamino)-3-(2-thienyl)-2-propenoic acid

Hazard: Irritant

Formula: C₉H₉NO₃S
MDL Number: MFCD01313644
CAS Number: 68762-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR