1,3-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]benzene ≥90%
Supplier: Matrix Scientific
115249-500MG
115249-1G
102899-160EA
372.31
USD
102899-160
102899-162
1,3-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]benzene ≥90%
1,3-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]benzene
Hazard: Irritant
Formula:
C₁₈H₃₄O₂Si₂ MW: 338.64 g/mol |
MDL Number:
MFCD28023550 CAS Number: 120951-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...