(S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane bis(borane) ≥95%
Supplier: Matrix Scientific
Synonyms:
(S,S)-1,2-Bis[boranato(tert-butyl)methylphosphino]ethane, Borane - (S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane Complex
115245-500MG
115245-1G
102895-676EA
1732.31
USD
102895-676
102895-732
(S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane bis(borane) ≥95%
(S,S)-1,2-Bis[(tert-butyl)methylphosphino]ethane bis(borane)
Hazard: Irritant
Formula:
C₁₂H₃₄B₂P₂ MW: 261.97 g/mol |
MDL Number:
MFCD03844770 CAS Number: 203000-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...